• USA Home
  • M7189 - Maltosyl β-cyclodexrin mixture (mono-/di-)

EMAIL THIS PAGE TO A FRIEND
M7189 Sigma

Maltosyl β-cyclodexrin mixture (mono-/di-)

solid

Purchase

Properties

form   solid
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?