• USA Home
  • M7259 - 4-Methylumbelliferyl palmitate

EMAIL THIS PAGE TO A FRIEND
M7259 Sigma

4-Methylumbelliferyl palmitate

Synonym: Palmitic acid 4-methylumbelliferyl ester

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Enzyme Substrate, Fatty Acyls, Fluorescent Enzyme Substrates,
InChI Key   QKQVFFACWLFRRX-UHFFFAOYSA-N
mp   77 °C(lit.)
fluorescence   λex 360 nm; λem 449 nm in 0.1 M phosphate pH 8.0
storage temp.   −20°C

Description

Application

Fluorogenic substrate for acid lipase in human leukocytes.

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?