• USA Home
  • M7266 - Myosin, Calcium activated from chicken muscle

EMAIL THIS PAGE TO A FRIEND
M7266 Sigma

Myosin, Calcium activated from chicken muscle

0.5-1.5 units/mg protein (biuret)

Purchase

Properties

Related Categories Biochemicals and Reagents, Myosin, Proteins and Derivatives, Structural Proteins
form   buffered aqueous glycerol solution
mol wt   heavy chain mol wt ~200 kDa (each)
  light chain mol wt 15-20 kDa (each)
  mol wt ~480 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   chicken ... MYH2(374069), MYH4(417308), MYL1(396470)

Description

Unit Definition

One unit will liberate 1.0 μmole of inorganic phosphorus from ATP per min at pH 9.0 at 25 °C in the presence of calcium.

Physical form

Solution in 50% glycerol containing 0.6 M KCl and 10 mM potassium phosphate buffer, pH 6.8.

Biochem/physiol Actions

Myosin interacts with actin in muscle and non-muscle cells. Myosin molecules consist of two major regions: tails (rods) and heads. They aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Myosin molecules spontaneously assemble into filaments in solutions of physiologic ionic strength and pH. Thick filament consists mainly of myosin molecules. Myosin is activated by the enzyme ATPase. This activation is the immediate source of the free energy that drives muscle contraction. It binds to the polymerized form of actin, the major constituent of the thin filament. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle. Isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. These are designated as type I (slow-twitch) and type II (fast-twitch). Myosin is one of three classes of cytoskeletal motor proteins that have been identified. The others are kinesins and dyneins. These three types of proteins are thought to be responsible for the many movements that occur in cells. Myosin has been shown to be involved in neurosensory function, vesicle trafficking, determinant partitioning, and cortical function.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?