• USA Home
  • M7527 - Myoglobin from sperm whale skeletal muscle

EMAIL THIS PAGE TO A FRIEND
M7527 Sigma

Myoglobin from sperm whale skeletal muscle

recombinant, expressed in E. coli, 95-100%, buffered aqueous solution

Purchase

Properties

Related Categories Biochemicals and Reagents, Coagulation Proteins and Reagents, Plasma & Blood Proteins, Proteins and Derivatives
recombinant   expressed in E. coli
assay   95-100%
form   buffered aqueous solution
Iron content   ~0.30%
storage temp.   −20°C

Description

Other Notes

Contains an N-terminal methionine not present in the natural product. Amino acid sequence not confirmed by Sigma.

Physical form

Solution in 0.02 M Tris HCl, pH 8.0

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Proteins on Discovery® BIO PolyMA-WAX (Analyte Set #1)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Proteins on Discovery® BIO PolyMA-WAX (Analyte Set #1)
Keywords: Chromatography, High performance liquid chromatography, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?