• USA Home
  • M7559 - Macrophage Colony Stimulating Factor Receptor/Fc Chimera human

EMAIL THIS PAGE TO A FRIEND
M7559 Sigma

Macrophage Colony Stimulating Factor Receptor/Fc Chimera human

>97% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym: CD115, M-CSF Receptor

Purchase

Properties

Related Categories Cell Biology, Cell Culture, Cell Signaling and Neuroscience, Cytokines and Growth Factors, Cytokines, Growth Factors and Hormones,
recombinant   expressed in NSO cells
assay   >97% (SDS-PAGE)
form   lyophilized powder
impurities   endotoxin, tested
storage temp.   −20°C
Gene Information   human ... CSF1R(1436)

Description

Analysis Note

The biological activity is measured by its ability to inhibit the M-CSF induced proliferation of M-NFS-60 cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 5.0 mg bovine serum albumin.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?