EMAIL THIS PAGE TO A FRIEND
M7750 Sigma

Mercury Orange

powder

Synonym: 1-(4-Chloromercuriophenylazo)-2-naphthol

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, M, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   CDEIGFNQWMSEKG-QIKYXUGXSA-M
form   powder
color   dark red

Description

Packaging

100 mg in glass bottle

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?