• USA Home
  • M7777 - Polaroid® MP4+ macro extension
EMAIL THIS PAGE TO A FRIEND
M7777 Sigma
Polaroid® MP4+ macro extension
Macro extension
Purchase
Properties
Related Categories Electrophoresis Camera Equipment, Electrophoresis Photography Supplies, Electrophoresis Supplies, Labware
availability   not available in EU
Description
Other Notes
For sliding camera head to increase magnification range and working distance of camera. With the macro extension, lenses have magnification ranges as follows: 135mm (up to 3.0×); 105mm (up to 4.0×); 75mm (1.5× to 6.5×); 35mm (5× to 14.3×)
Legal Information
Polaroid is a registered trademark of Polaroid Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?