• USA Home
  • M8007 - Myoglobin from canine heart

EMAIL THIS PAGE TO A FRIEND
M8007 Sigma

Myoglobin from canine heart

95-100%, essentially salt-free, lyophilized powder

Purchase

Properties

assay   95-100%
form   essentially salt-free, lyophilized powder
purified by   crystallization
Iron content   ~0.20%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

M1882 Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
M0630 Myoglobin from equine skeletal muscle, 95-100%, essentially salt-free, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?