EMAIL THIS PAGE TO A FRIEND
M8021 Sigma

N-Methyl-L-proline

  • CAS Number 475-11-6

  • Empirical Formula (Hill Notation) C6H11NO2

  • Molecular Weight 129.16

  •  MDL number MFCD00011565

  •  PubChem Substance ID 24897214

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   CWLQUGTUXBXTLF-YFKPBYRVSA-N
storage temp.   −20°C

Description

Biochem/physiol Actions

N-Methyl-L-proline is a proline analogue present in citrus fruits.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?