EMAIL THIS PAGE TO A FRIEND
M8034 Sigma
Semi-dry blotter
Replacement Mylar® shield
Purchase
Properties
Related Categories Blotters, Electrophoresis Supplies, Labware, Semi-Dry Blotters
Description
General description
Excellent for Western, Northern, or Southern transfers, unit accepts one or several gels up to 20 × 20 cm, and varying thickness.
Stainless steel cathode and platinum anode plates produce highly uniform field and resist corrosion. Mylar shield provided to mask unused area of cathode, prevents arcing, ensures all current passes through gel.
Mylar shield masks unused area of cathode, ensuring that all current passes through gel.
Legal Information
Mylar is a registered trademark of Dupont Teijin Films
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?