• USA Home
  • M8038 - Multiple Antigenic Peptide (MAP) Core-PAM Resin

EMAIL THIS PAGE TO A FRIEND
M8038 Sigma

Multiple Antigenic Peptide (MAP) Core-PAM Resin

Synonym: Boc8-Lys4-Lys2-Lys-β-Ala Resin

Purchase

Properties

extent of labeling   0.8 mmol per g resin

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?