• USA Home
  • M8059 - [Ala107]-Myelin Basic Protein Fragment 104-118

EMAIL THIS PAGE TO A FRIEND
M8059 Sigma

[Ala107]-Myelin Basic Protein Fragment 104-118

Synonym: [Ala107]-MBP fragment 104-118

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Kinase/Phosphatase Biology, Myelin-Associated Glycoprotein,
InChI Key   PKKMMDDHSZRJTF-NBRSAOMZSA-N
storage temp.   −20°C
Gene Information   human ... MBP(4155)

Description

Amino Acid Sequence

Gly-Lys-Gly-Ala-Gly-Leu-Ser-Leu-Ser-Arg-Phe-Ser-Trp-Gly-Ala

Biochem/physiol Actions

[Ala107]-Myelin Basic Protein Fragment 104-118 is an inhibitor of protein kinase C.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?