• USA Home
  • M8155 - [Ala8,13,18]-Magainin II amide

EMAIL THIS PAGE TO A FRIEND
M8155 Sigma

[Ala8,13,18]-Magainin II amide

≥97% (HPLC)

Purchase

Description

Amino Acid Sequence

Gly-Ile-Gly-Lys-Phe-Leu-His-Ala-Ala-Lys-Lys-Phe-Ala-Lys-Ala-Phe-Val-Ala-Glu-Ile-Met-Asn-Ser-NH2

Biochem/physiol Actions

Magainin analog with greater anti-microbial activity.

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?