• USA Home
  • M8184 - Myelin Basic Protein Fragment 104-118

EMAIL THIS PAGE TO A FRIEND
M8184 Sigma

Myelin Basic Protein Fragment 104-118

synthetic, lyophilized powder

Synonym: MBP (104-118)

Purchase

Description

Amino Acid Sequence

Gly-Lys-Gly-Arg-Gly-Leu-Ser-Leu-Ser-Arg-Phe-Ser-Trp-Gly-Ala

Biochem/physiol Actions

Substrate of Protein Kinase C

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?