• USA Home
  • M8187 - Greiner Multiwell Strips
EMAIL THIS PAGE TO A FRIEND
M8187 Sigma
Greiner Multiwell Strips
96 well plate, medium binding, black flat bottom (1x8 well strips)
Synonym: Greiner Multiwell Strips
Purchase
Properties
Related Categories 96 Well Plates, Greiner 96-Well Multiwell Plates, Labware, Multiwell Plates
material   black flat bottom (1x8 well strips)
mfr. no.   Greiner 762076
size   96 wells
Description
General description
Both medium-binding and high binding are offered. High binding plates and strips are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding are suggested.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?