EMAIL THIS PAGE TO A FRIEND
M8418 Sigma

MAN-3 Glycan

≥80% (MS)

Synonym: (Man)3(GlcNAc)2, Mannotriose-di-(N-acetyl-D-glucosamine)

  • CAS Number 70858-45-6

  • Empirical Formula (Hill Notation) C34H58N2O26

  • Molecular Weight 910.82

  •  MDL number MFCD00467028

  •  PubChem Substance ID 24897254

Purchase

Properties

Related Categories Glycan Component Classes, Glycobiology, Molecular Biology, N-Glycans, N-Linked High Mannose Glycans,
InChI Key   DJUIFTOHRFYXSA-UHFFFAOYSA-N
assay   ≥80% (MS)
storage temp.   2-8°C

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

N-Linked Glycans

N-linked glycosylation, modification, and degradation are involved in a wide variety of processes in all organisms from archaea to eukaryotes. It is the most common covalent protein modification in e...
Glycobiology Analysis Manual, 2nd Edition
Keywords: Catalysis, Degradations, Diseases, Eliminations, Fucosylation, Glycosylations, Metabolism, N-linked glycosylation, Sequences, Substitutions, Type, transformation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?