• USA Home
  • M8420 - MECC buffer solution A
EMAIL THIS PAGE TO A FRIEND
M8420 Sigma
MECC buffer solution A
suitable for separation of water-soluble vitamins with CE
Purchase
Properties
sterility   0.2 μm filtered
suitability   suitable for separation of water-soluble vitamins with CE
storage temp.   2-8°C
Description
Other Notes
20 mM sodium borate/phosphate, pH 9.0, and 50 mM SDS.
Price and Availability
Safety & Documentation
Safety Information
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?