• USA Home
  • M8430 - Anti-Matrix Metalloproteinase-8, Propeptide Region antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
M8430 Sigma

Anti-Matrix Metalloproteinase-8, Propeptide Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-MMP-8

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Extracellular Matrix, Antibodies to Matrix Metalloproteinases and TIMPs,
species reactivity   human
application(s)   western blot (chemiluminescent): 1:5,000
  western blot: 1:1,000
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
mol wt   antigen mol wt 85 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... MMP8(4317)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the propeptide region of human matrix metalloproteinase-8 (neutrophil collagenase, collagenase-2).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide.

Specificity

Reacts with reduced and non-reduced latent MMP-8. The propeptide region is shed after activation; therefore, the antibody recognizes only the latent form of MMP-8. May be used to distinguish between latent and active MMP-8.

Price and Availability


Antibody Explorer

Explore with Confidence

Biomedical Applications
Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Safety Information

WGK Germany  1
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?