EMAIL THIS PAGE TO A FRIEND
M8546 Sigma

Mannide monooleate

liquid

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Polysaccharide More...
InChI Key   NWGKJDSIEKMTRX-AAZCQSIUSA-N
form   liquid
color   yellow

Description

Application

A non-ionic surface active emulsifier.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M8819 Mannide monooleate, from plant

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?