• USA Home
  • M8656 - Methyl 2,3,4-tri-O-acetyl-β-D-thiogalactopyranosiduronic acid methyl ester

EMAIL THIS PAGE TO A FRIEND
M8656 Sigma

Methyl 2,3,4-tri-O-acetyl-β-D-thiogalactopyranosiduronic acid methyl ester

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates M-O, Monosaccharide More...
InChI Key   HHAWIPPOHLEARU-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?