• USA Home
  • M8685 - Minimum Essential Medium Eagle

EMAIL THIS PAGE TO A FRIEND
M8685 Sigma

Minimum Essential Medium Eagle

Alpha Modification, with L-glutamine, sodium bicarbonate, ribonucleosides, and deoxyribonucleosides, liquid, sterile-filtered, cell culture tested

Synonym: MEM

Purchase

Properties

sterility   sterile-filtered
form   liquid
IVD   for in vitro diagnostic use
impurities   endotoxin, tested
suitability   cell culture tested
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M8042 Minimum Essential Medium Eagle, Alpha Modification, with ribonucleosides, deoxyribonucleosides and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?