• USA Home
  • M8894 - IgG2b, Kappa from murine myeloma

EMAIL THIS PAGE TO A FRIEND
M8894 Sigma

IgG2b, Kappa from murine myeloma

clone MOPC 141, purified immunoglobulin, buffered aqueous solution

Synonym: Mouse IgG2b-κ

Purchase

Properties

Related Categories Antibodies, Mouse Myeloma Proteins, Purified Immunoglobulins, Sera and Immunoglobulins, Supplementary Products More...
biological source   mouse
antibody form   purified immunoglobulin
clone   MOPC 141, monoclonal
form   buffered aqueous solution
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Physical form

Solution in 0.02 M Tris buffered saline, pH 8.0, containing 0.02% sodium azide

Application

Monoclonal clone MOPC 141 IgG2b, Kappa light chain may be used as an antigen for the production and evaluation of antibodies developed for the indentification and classification of IgG2 subtypes.

General description

Mammalian immunoglobins contain either lambda or kappa light chains.

Specificity of the mouse IgG2b, Kappa, is determined by mouse monoclonal isotyping strips. The purified immunoglobulin preparation is non-reactive with anti-mouse IgA, IgM, IgG1, IgG2a and IgG3.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?