• USA Home
  • M8939 - Thumbs-Up microtubes
EMAIL THIS PAGE TO A FRIEND
M8939 Sigma
Thumbs-Up microtubes
volume 0.5 mL
Purchase
Description
General description
Saves fingernails from breaking and cracking. Opens with a flip of a thumb. Reduces sample contamination. Fast and easy removal from Rotors. Inner lid structure allows for pre-loading of reactants. RNase and DNase free. Fully autoclavable propylene. RCF rating of 17,000 × g. Can withstand temperatures ranging from - 196°C to 130°C.
Legal Information
Thumbs-Up is a trademark of Diversified Biotech
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?