EMAIL THIS PAGE TO A FRIEND
M9013 Sigma

2-O-Methyl-D-xylose

  • CAS Number 7434-28-8

  • Empirical Formula (Hill Notation) C6H12O5

  • Molecular Weight 164.16

  •  PubChem Substance ID 329818076

Purchase

Properties

InChI Key   ALNDFFUAQIVVPG-UHFFFAOYSA-N

Description

Other Notes

A mixture of α- and β-isomers.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?