• USA Home
  • M9046 - 1-Methylpiperidine solution

EMAIL THIS PAGE TO A FRIEND
M9046 Sigma

1-Methylpiperidine solution

15% in methanol: water

Synonym: N-Methylpiperidine

  • CAS Number 626-67-5

  • Empirical Formula (Hill Notation) C6H13N

  • Molecular Weight 99.17

  •  Beilstein Registry Number 1560282

  •  MDL number MFCD00006491

  •  PubChem Substance ID 329818079

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   PAMIQIKDUOTOBW-UHFFFAOYSA-N
concentration   15% in methanol: water
storage temp.   2-8°C

Description

Caution

Shake well prior to use.

Packaging

20% price reduction on quantities of 4 or more

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3286 3/PG 2
WGK Germany 
3
Flash Point(F) 
41 °F
Flash Point(C) 
5 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?