• USA Home
  • M9388 - 2-(Methylamino)ethanol

EMAIL THIS PAGE TO A FRIEND
M9388 Sigma

2-(Methylamino)ethanol

Synonym: N-Methylethanolamine

Purchase

Properties

density   0.94 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Epididymus, Kidney, Liver, Testes, ovary, spleen
RIDADR 
UN 2735PSN1 8 / PGII
WGK Germany 
1
RTECS 
KL6650000
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?