• USA Home
  • M9638 - γ-Melanocyte Stimulating Hormone

EMAIL THIS PAGE TO A FRIEND
M9638 Sigma

γ-Melanocyte Stimulating Hormone

≥97% (HPLC)

Synonym: γ-MSH

  • CAS Number 72711-43-4

  • Empirical Formula (Hill Notation) C74H99N21O16S

  • Molecular Weight 1570.77

  •  MDL number MFCD00133358

  •  PubChem Substance ID 329818095

Purchase

Description

Amino Acid Sequence

Tyr-Val-Met-Gly-His-Phe-Arg-Trp-Asp-Arg-Phe-Gly

Linkage

Derived from β-lipotropin (45-56)

Biochem/physiol Actions

Hormone that stimulates melanogenesis; facilitates learning and memory; affects inflammatory and immune responses and peripheral nerve regeneration.

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?