• USA Home
  • M9660 - Ultrafree-MC microcentrifuge filters
EMAIL THIS PAGE TO A FRIEND
M9660 Sigma
Ultrafree-MC microcentrifuge filters
capacity 500 μL, Durapore PVDF membrane, pore size 0.45 μm
Purchase
Properties
Related Categories Centrifuge Filtration, Filtration Supplies, Labware, Manufacturer Browser, Millipore,
material   Durapore PVDF membrane
mfr. no.   Millipore, UFC30HV25
limit   12,000 × g max. RCF
capacity   500 μL
pore size   0.45 μm
Description
Compatibility
Use in any standard fixed-angle microcentrifuge rotor.
General description
Units consist of microcentrifuge tube with hinged cap and filter insert. The Ultrafree centrifugal filter units are are well suited for the removal of particles & precipitates from aqueous & some solvent-based samples.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?