Synonym: Ribothymidine
CAS Number 1463-10-1
Empirical Formula (Hill Notation) C10H14N2O6
Molecular Weight 258.23
EC Number 215-973-9
MDL number MFCD00006535
Popular Documents: FTNMR (PDF)
| mp | 183-184 °C(lit.) |
| storage temp. | 2-8°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?