• USA Home
  • M9924 - Anti-Mouse IgG3 (heavy chain specific) antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
M9924 Sigma

Anti-Mouse IgG3 (heavy chain specific) antibody produced in goat

fractionated antiserum, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, MO-MR, Primary Antibodies
biological source   goat
antibody form   fractionated antiserum
clone   polyclonal
form   lyophilized powder
conjugate   unconjugated
storage temp.   2-8°C

Description

Reconstitution

Reconstitute with 2 ml deionized water.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

comparable product

Product #

Description

Add to Cart

M8270 Anti-Mouse IgG3 (heavy chain specific) antibody produced in goat, affinity isolated antibody, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?