• USA Home
  • M9927 - McBride listeria agar base

EMAIL THIS PAGE TO A FRIEND
M9927 Sigma

McBride listeria agar base

pH 5.5±0.2 (25 °C)

Purchase

Properties

pH   5.5±0.2 (25 °C)

Description

Application

Used for the selective isolation and cultivation of Listeria monocytogenes from foods and clinical samples.

Other Notes

Components (g/L)
Tryptose 10.00
Beef extract 3.00
Sodium chloride 5.00
Glycine anhydride 10.00
Lithium chloride 0.50
Phenyl ethanol 2.50
Agar 15.00

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?