• USA Home
  • MBK1 - Maleimide Activated BSA, KLH Conjugation Kit

EMAIL THIS PAGE TO A FRIEND
MBK1 Sigma

Maleimide Activated BSA, KLH Conjugation Kit

Purchase

Description

Caution

Alkaline pH values (above 7.5) may hydrolyze the maleimide group or generate side reactions with amines. Haptens must contain cysteine or a sulfhydryl group in the reduced state in order to react efficiently with the maleimide group.

Features and Benefits

• Conjugation and purification in under 3 hours
• BSA and KLH are pre-activated and ready to use
• Mild reaction conditions: pH 6.5-7.5 in aqueous buffer
• Fast gel filtration separation of conjugate and free hapten
• Prepare immunogen and antigen for immunoassay with one kit
• Contains sufficient reagents for 3 conjugations each of BSA and KLH. Each reaction can conjugate at least 4 mg of hapten.
• Contains reagents for coupling efficiency assay
• Complete protocols and references included

Principle

Sigma offers a kit for conjugating haptens to bovine serum albumin (BSA) and keyhole limpet hemocyanin (KLH). BSA and KLH have been used as protein carriers for haptens in immunization and immunoassay protocols. Immunogen is prepared with one carrier protein, typically KLH, while the antigen for immunoassay is prepared with another carrier, typically BSA. This eliminates the need to remove antibodies developed against the immunogen carrier protein from the antiserum before assay.

Specificity

KLH and BSA have been activated with succinimidyl-3-N-maleimidopropionic acid to provide a maleimide group at the end of a three atom spacer. Under approximately neutral conditions (pH 6.5-7.5) the maleimide will form a thioether bond with a sulfhydryl group, i.e., a cysteine residue on a peptide.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection

Kit component also available separately

Description

Product #

Add to Cart

Maleimide Activated BSA 3 x 5 mg SDS B7542
Maleimide Activated KLH 3 x 5 mg SDS K0383
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

F5506 Freund’s Adjuvant, Incomplete, liquid
H4397 TiterMax® Classic Adjuvant, liquid
T2684 TiterMax® Gold Adjuvant, liquid
F5881 Freund’s Adjuvant, Complete, cell suspension

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?