• USA Home
  • MWSDS70L - Molecular Weight Marker Kit

EMAIL THIS PAGE TO A FRIEND
MWSDS70L Sigma

Molecular Weight Marker Kit

lyophilized powder, for Molecular Weights 14,000-66,000

Purchase

Properties

form   lyophilized powder
storage temp.   2-8°C

Description

Packaging

This kit contains one vial each of seven individual proteins, one vial of the standard mixture (SDS-7) and a technical bulletin.

Reconstitution

The individual markers must be reconstituted with sample buffer (S 3401) at a recommended concentration of 1 mg/ml.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

S3401 Sample Buffer, Laemmli 2× Concentrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?