• USA Home
  • N0139 - Nitrocellulose membranes for hybridization

EMAIL THIS PAGE TO A FRIEND
N0139 Sigma

Nitrocellulose membranes for hybridization

pore size 0.45 μm, size 33 cm × 56 cm

Purchase

Properties

Related Categories Hybridization Membranes, Hybridization Supplies, Labware, Molecular Biology Supplies
size   33 cm × 56 cm
pore size   0.45 μm

Description

Application

For use in dot blots and Southern blots

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3270 4.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

Z670944 Whatman® Protran® nitrocellulose membranes, Sheets - BA85, pore size 0.45 μm, L × W 33 cm × 56 cm

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?