• USA Home
  • N0519 - Nitrate Reductase (cytochrome) from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
N0519 Sigma

Nitrate Reductase (cytochrome) from Escherichia coli

lyophilized powder, ≥5 units/g solid

Synonym: Ferrocytochrome:Nitrate oxidoreductase

Purchase

Description

Unit Definition

One unit will reduce 1.0 μmole of nitrate to nitrite per min at pH 7.0 at 30 °C in a methylviologen system.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?