EMAIL THIS PAGE TO A FRIEND
N0626 Sigma

1-Naphthyl stearate

Synonym: Octadecanoic acid 1-naphthyl ester

Purchase

Properties

Related Categories Activity, Activity Other, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates More...
InChI Key   CJVDJPQHLSHHMZ-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?