• USA Home
  • N0788 - N-TER Nanoparticle siRNA Transfection System

EMAIL THIS PAGE TO A FRIEND
N0788 Sigma

N-TER Nanoparticle siRNA Transfection System

Purchase

Properties

Related Categories Functional Genomics and RNAi, MISSION siRNA, Molecular Biology, N-TER Nanoparticle siRNA Transfection System
shipped in   wet ice
storage temp.   −20°C

Description

Legal Information

Manufactured and Distributed by Sigma-Aldrich, under license from the CNRS.

N-TER is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Kit component only

Description

Product #

Add to Cart

N-TER Peptide SDS    
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

N2913 N-TER Nanoparticle siRNA Transfection System

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?