• USA Home
  • N0909 - Nociceptin Fragment 1-7

EMAIL THIS PAGE TO A FRIEND
N0909 Sigma

Nociceptin Fragment 1-7

Synonym: Orphanin FQ Fragment 1-7

  • Empirical Formula (Hill Notation) C31H41N7O9

  • Molecular Weight 655.70

  •  MDL number MFCD02684529

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
storage temp.   −20°C
Gene Information   human ... PNOC(5368)

Description

Amino Acid Sequence

Phe-Gly-Gly-Phe-Thr-Gly-Ala

Biochem/physiol Actions

Reduces nociceptin-induced hyperalgesia in mice. This endogenously formed N-terminal fragment may modulate the hyperalgesic action of nociceptin in the spinal cord.

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?