• USA Home
  • N1035 - N-(NBD-aminododecanoyl)-L-α-phosphatidyl-DL-glycerol, α-oleoyl sodium salt

EMAIL THIS PAGE TO A FRIEND
N1035 Sigma

N-(NBD-aminododecanoyl)-L-α-phosphatidyl-DL-glycerol, α-oleoyl sodium salt

≥95%, solid

Synonym: 1-(cis-9-Octadecenoyl)-2-[12-[(7-nitro-2-,1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-sn-glycerol-3-phospho-rac-(1-glycerol) sodium salt, 18:1-12:0 NBD PG

  • Empirical Formula (Hill Notation) C42H70N4NaO13P

  • Molecular Weight 892.99

  •  MDL number MFCD03453618

Purchase

Description

Other Notes

Fluorescently labeled fatty acid phospholipid membrane probe

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?