• USA Home
  • N1141 - 19-Nor-4-androstene-3,17-dione

EMAIL THIS PAGE TO A FRIEND
N1141 Sigma

19-Nor-4-androstene-3,17-dione

Synonym: 4-Estrene-3,17-dione

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Biomarkers and Metabolomics Standards, Hormone Standards
InChI Key   JRIZOGLBRPZBLQ-QXUSFIETSA-N
form   neat
drug control   USDEA Schedule IIIN; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?