• USA Home
  • N1147 - Neuropeptide Y fragment

EMAIL THIS PAGE TO A FRIEND
N1147 Sigma

Neuropeptide Y fragment

≥97% (HPLC)

Purchase

Properties

assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?