EMAIL THIS PAGE TO A FRIEND
N1269 Sigma

19-Nortestosterone

1.0 mg/mL in 1,2-dimethoxyethane

Synonym: 17β-Hydroxy-19-norandrost-4-en-3-one, 17β-Hydroxy-4-estren-3-one, 4-Estren-17β-ol-3-one, Nandrolone, Norandrostenolone

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Biomarkers and Metabolomics Standards, Hormone Standards
drug control   USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in 1,2-dimethoxyethane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2252 3 / PGII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?