• USA Home
  • N1272 - Neuropeptide Y Fragment 18-36 human, porcine

EMAIL THIS PAGE TO A FRIEND
N1272 Sigma

Neuropeptide Y Fragment 18-36 human, porcine

Synonym: NPY 18-36

  • CAS Number 98264-90-5

  • Empirical Formula (Hill Notation) C112H174N36O27

  • Molecular Weight 2456.80

  •  MDL number MFCD00214635

  •  PubChem Substance ID 329818547

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Gastrointestinal Peptides, Neuropeptide Y, Neuropeptide Y and Peptide YY Peptides,
InChI Key   KPPFDQVOSMXZBV-VMRDKNQRSA-N
assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C
Gene Information   human ... NPY(4852)
pig ... NPY(397304)

Description

Amino Acid Sequence

Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2

Biochem/physiol Actions

A competitive NPY antagonist in cardiac membranes

Price and Availability


Making Headway

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?