• USA Home
  • N1382 - 4-Nitrophenyl β-D-thiofucopyranoside

EMAIL THIS PAGE TO A FRIEND
N1382 Sigma

4-Nitrophenyl β-D-thiofucopyranoside

  • CAS Number 51885-02-0

  • Empirical Formula (Hill Notation) C12H15NO6S

  • Molecular Weight 301.32

  •  PubChem Substance ID 329818574

Purchase

Properties

InChI Key   JVQFCJCGZQPUHG-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?