• USA Home
  • N1389 - Nylon membranes, charge modified for hybridization
EMAIL THIS PAGE TO A FRIEND
N1389 Sigma
Nylon membranes, charge modified for hybridization
pore size 0.45 μm, size 20 cm × 20 cm
Purchase
Properties
Related Categories Core Bioreagents, Hybridization Membranes, Hybridization Supplies, Labware, Life Science Reagents for Northern and Southern Blotting,
size   20 cm × 20 cm
pore size   0.45 μm
suitability   suitable for electroblotting of nucleic acids down to approx. 100 base pairs
Description
Other Notes
Reusable, non-curling, positively charged Nylon 66 membranes
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?