EMAIL THIS PAGE TO A FRIEND
N138 Sigma

(+)-Normetazocine

Synonym: (+)-(2S,[2α,6α,11R])-1,2,3,4,5,6-Hexahydro-6,11-dimethyl-2,6-methano-3-benzazocin-8-ol

  • CAS Number 16670-83-0

  • Empirical Formula (Hill Notation) C14H19NO

  • Molecular Weight 217.31

  •  MDL number MFCD00167442

Purchase

Properties

InChI Key   DXESFJJJWBHLJX-IIMNLJJBSA-N
form   neat
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?