• USA Home
  • N140 - N-(p-Isothiocyanatophenethyl)spiperone hydrochloride

EMAIL THIS PAGE TO A FRIEND
N140 Sigma

N-(p-Isothiocyanatophenethyl)spiperone hydrochloride

Synonym: NIPS hydrochloride

  • CAS Number 135261-88-0

  • Empirical Formula (Hill Notation) C32H33FN4O2S · HCl

  • Molecular Weight 593.15

  •  MDL number MFCD00153849

  •  PubChem Substance ID 24897486

Purchase

Properties

Related Categories Antagonists, Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Dopaminargics - Antagonists,
InChI Key   MHDTWYNWOBNGFA-UHFFFAOYSA-N
color   light tan
solubility   DMSO: soluble
  aqueous base: unstable
  ethanol: soluble
storage temp.   −20°C

Description

Biochem/physiol Actions

Selective irreversible D2 dopamine receptor antagonist.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?