• USA Home
  • N147 - [β-Ala8]-Neurokinin A Fragment 4-10

EMAIL THIS PAGE TO A FRIEND
N147 Sigma

[β-Ala8]-Neurokinin A Fragment 4-10

≥97% (HPLC), powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, GPCR Modulators, GPCR Proteins, Modulators and Antibodies,
InChI Key   CKNPSJOMUQBPLA-WTWMNNMUSA-N
assay   ≥97% (HPLC)
form   powder
composition   Peptide content, ≥60%
color   white
storage temp.   −20°C
Gene Information   human ... TAC1(6863)

Description

Amino Acid Sequence

Asp-Ser-Phe-Val-β-Ala-Leu-Met-NH2

Biochem/physiol Actions

Potent and selective NK-2 tachykinin receptor agonist.

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?