• USA Home
  • N157 - Naltrindole isothiocyanate hydrochloride

EMAIL THIS PAGE TO A FRIEND
N157 Sigma

Naltrindole isothiocyanate hydrochloride

solid

Synonym: NTII

  • CAS Number 126876-64-0

  • Empirical Formula (Hill Notation) C27H25N3O3S · HCl

  • Molecular Weight 508.03

  •  PubChem Substance ID 329818585

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience,
InChI Key   SYDXFYIGIIAPLB-OKRPGNDBSA-N
form   solid
color   off-white
solubility   45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: soluble3.3 mg/mL
  DMSO: soluble
  H2O: insoluble
  ethanol: slightly soluble
storage temp.   −20°C

Description

Biochem/physiol Actions

Selective irreversible δ2 opioid receptor antagonist.

Packaging

5 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?