• USA Home
  • N1768 - N-Nitrosodiisopropanolamine

EMAIL THIS PAGE TO A FRIEND
N1768 Sigma

N-Nitrosodiisopropanolamine

viscous liquid

Purchase

Properties

InChI Key   MNIGYIKCFSPQRJ-UHFFFAOYSA-N
form   viscous liquid
impurities   ≤10% Isopropanol (actual content given on label)
color   yellow
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
JL9650000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?